Anti-SLC45A3 Rabbit Polyclonal Antibody
PRSI47-137
- Antigen name:Solute Carrier Family 45 Member A3
- Antigen symbol:SLC45A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:IPCA6|PCANAP8|IPCA-2|IPCA-6|PCANAP6|PCANAP2|PRST|IPCA-8
- Immunogen:Synthetic peptide corresponding to aa residues 11-28 of human EAPII conjugated to KLH at the C-terminus.
- Tested applications:Immunoaffinity Chromatography
- Size:100 µg
- Pk:100 µG
ETS proteins constitute a family of conserved sequence-specific DNA-binding proteins and function as transcription factors with roles in differentiation, lymphoid cell develop-ment, invasiveness, and angiogenesis. A novel ETS1 interacting protein designated as ETS1-associated protein II (EAPII) interacts with ETS1 and other ETS proteins (ETS2 and FLI1) and is a novel ETS1 modulator.
PBS
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SLC45A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human