Order Entry
Poland
ContactUsLinkComponent
Anti-AS3MT Goat Polyclonal Antibody
Anti-AS3MT Goat Polyclonal Antibody
Catalog # PRSI45-299
Supplier:  ProSci Inc.
undefined
Anti-AS3MT Goat Polyclonal Antibody
Catalog # PRSI45-299
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Arsenite Methyltransferase
  • Antigen symbol:
    AS3MT
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    AS3MT
  • Concentration:
    500 ug/mL
  • Immunogen:
    AS3MT antibody was raised against a 13 amino acid synthetic peptide near the N-Terminus of AS3MT.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

AS3MT antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

AS3MT antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: AS3MT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: