Order Entry
Poland
ContactUsLinkComponent
Anti-LMAN2 Rabbit Polyclonal Antibody
Anti-LMAN2 Rabbit Polyclonal Antibody
  PRSI30-229
undefined
Anti-LMAN2 Rabbit Polyclonal Antibody
  PRSI30-229

 

  • Antigen name:
    Lectin Mannose-Binding 2
  • Antigen symbol:
    LMAN2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Dog,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    Chromosome 5 Open Reading Frame 8|Vesicular Integral Protein Of 36 Kda|GP36B|VIP36|Vesicular Integral-Membrane Protein 36
  • Immunogen:
    A synthetic peptide corresponding a region of human LMAN2.
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

LMAN2 plays a role as an intracellular lectin in the early secretory pathway. It interacts with N-acetyl-D-galactosamine and high-mannose type glycans and may also bind to O-linked glycans. It is involved in the transport and sorting of glycoproteins carrying high mannose-type glycans.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: LMAN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse