Order Entry
Poland
ContactUsLinkComponent
Anti-FOXE3 Rabbit Polyclonal Antibody
Anti-FOXE3 Rabbit Polyclonal Antibody
Catalog # PRSI27-546
Supplier:  ProSci Inc.
undefined
Anti-FOXE3 Rabbit Polyclonal Antibody
Catalog # PRSI27-546
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Forkhead Box E3
  • Antigen symbol:
    FOXE3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    FREAC8|FKHL12
  • Immunogen:
    A synthetic peptide corresponding a region of human FOXE3.
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Forkhead Box Protein E3 (FOXE3, forkhead-related protein FKHL12, forkhead-related transcription factor 8) is a forkhead/winged helix transcription factor, which is expressed in the developing lens from the start of lens placode induction and becomes restricted to the anterior proliferating cells when lens fiber differentiation begins.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FOXE3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human