Specifications
- Antigen name:Tripartite Motif Containing 28
- Antigen symbol:TRIM28
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:KRIP-1|TF1B|TIF1B|KAP-1|KAP1|MommeD9|PPP1R157|RNF96|AA408787
- Concentration:1 mg/mL
- Immunogen:Rabbit polyclonal TRIM28 antibody was raised against a 14 amino acid peptide near the carboxy terminus of human TRIM28.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
TRIM28 Antibody: TRIM28, also known as KAP-1, is a member of the Transcriptional Intermediary Factor 1 (TIF1) subfamily and contains a RING finger, B box, Coiled coil, PHD/TTC, and bromodomain. TRIM28 is a corepressor for Kruppel-associated box (KRAB)-domain-containing zinc finger proteins and plays a critical role in early embryogenesis. TRIM28 acts as a transcriptional mediator by binding liganded nuclear receptors, including retinoic acid (RAR), retinoid X (RXR) and estrogen (ER) receptors. TRIM28 associates with both heterochromatin and euchromatin, causing gene silencing by both HP1 binding and histone deacetylation.
TRIM28 antibody can be used for detection of TRIM28 by Western blot at 1 - 2 ?g/mL.
TRIM28 Antibody is affinity chromatography purified via peptide column.
TRIM28 Antibody is supplied in PBS containing 0.02% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TRIM28
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat