Anti-RILP Rabbit Polyclonal Antibody
Catalog # PRSI6791
Dostawca: ProSci Inc.
Dane techniczne
- Antigen name:Rab interacting lysosomal Protein
- Antigen symbol:RILP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Antigen synonyms:Rab-interacting lysosomal protein|PP10141|RILP
- Storage buffer:PBS containing 0.02% sodium azide
- Immunogen:RILP antibody was raised against an 18 amino acid synthetic peptide near the center of human RILP.
- Tested applications:E, WB, ICC, IF
- Purification:Affinity chromatography purified via peptide column
- Size:100 µg
- Pk:100 µG
Dane techniczne
Informacje o tym produkcie
RILP Antibody: The Rab interacting lysosomal protein (RILP) is lysosomal protein that interacts with RAB7, a small GTPase that controls transport to endocytic degradative compartments. It is thought that RILP is a downstream effector of RAB7 and both proteins act together in the regulation of late endocytic traffic. Overexpression of RILP caused enlarged lysosomes that are more centrally located in the cell suggesting that RILP may also be involved in the regulation of lysosomal morphology. Other evidence suggests that RILP may play a role in the biogenesis of multivesicular bodies.
Type:
Antigen: RILP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse