Order Entry
Poland
ContactUsLinkComponent
 
Anti-VPS53 Rabbit Polyclonal Antibody
Catalog # PRSI4593
Supplier:  ProSci Inc.
undefined
Anti-VPS53 Rabbit Polyclonal Antibody
Catalog # PRSI4593
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Vacuolar Protein Sorting 53
  • Antigen symbol:
    VPS53
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Antigen synonyms:
    HCCS1|Vacuolar protein sorting-associated protein 53 homolog|hVps53L|pp13624|VPS53
  • Storage buffer:
    PBS containing 0.02% sodium azide
  • Immunogen:
    VPS53 antibody was raised against a 18 amino acid synthetic peptide from near the carboxy terminus of human VPS53.
  • Tested applications:
    E, WB, ICC, IF
  • Purification:
    Affinity chromatography purified via peptide column
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

VPS53 Antibody: The sorting of acid hydrolases to lysosomes rely on mannose 6-phosphate receptors that cycle between the trans-Golgi network (TGN) and endosomes. The maintenance of this cycle requires the function of the mammalian Golgi-associated retrograde protein (GARP) complex which is composed of three subunits: VPS52, VPS53, and VPS54. Depletion of any of these three proteins, such as by RNAi, impairs the retrograde transport of multiple TGN proteins. VPS53 was identified as an HIV dependency factor (HDF) and plays a role in viral entry to the cell, suggesting that VPS53 may be an important drug target in HIV treatment. At least five isoforms of VPS53 are known to exist.

Type:
Antigen: VPS53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat