Order Entry
Poland
Orders LinkContactUsLinkComponent
Lactobionic acid 97%

Lactobionic acid 97%

Lactobionic acid 97%
Certificates
Lactobionic acid 97%

3 Options Available Below

Product Details & Documents

Soluble in water

Synonyms:  (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoic acid, 4-O-β-D-Galactopyranosyl-D-gluconic acid
  • CAS Number:96-82-2
  • Formula:C₁₂H₂₂O₁₂
  • MDL Number:MFCD00078147
  • Molecular Weight:358.30 g/mol
  • Merck Index:14,5342
  • Melting Pt:113…118 °C
  • Storage Temperature:Ambient
  • EINECS:202-538-3
Specifications for Product Family

Specification Test Results

  • Colour:
    White to Off-White
  • Form:
    Powder
  • Assay (Titration):
    97% min
  • Water Content (Karl Fischer Titration):
    5,0% max
Product Family Options

Product Information