Order Entry
Poland
ContactUsLinkComponent
Beclomethasone Dipropionate, Reference Standards
Beclomethasone Dipropionate, Reference Standards
Supplier:  USP
Certificates
Beclomethasone Dipropionate, Reference Standards
Supplier:  USP
Certificates

Specifications

  • CAS No.:
    5534-09-8
  • Formula:
    C₂₈H₃₇ClO₇
  • Description:
    Beclomethasone Dipropionate
  • Storage conditions:
    Ambient
  • Synonym(s):
    Beclomethasone-21-propionate; 9-Chloro-11β,17-dihydroxy-16β-methyl-3,20-dioxopregna-1,4-diene-21-yl propionate
  • Melting point:
    > 190 °C
  • Physical form:
    Solid powder
  • Boiling point:
    > 421 °C

Specifications

Related Information

Frequently Bought Together