Order Entry
Poland
Orders LinkContactUsLinkComponent
Phenylephrine Related Compounds, Reference Standard

Phenylephrine Related Compounds, Reference Standard

Supplier:   USP
Phenylephrine Related Compounds, Reference Standard
Supplier:  USP
Certificates
Phenylephrine Related Compounds, Reference Standard

5 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1533024.
  • 1533035.
  • 1533046.
  • 1533261.
  • 1533272.
  • Description:
  • Phenylephrine Related Compound C
  • Phenylephrine Related Compound D
  • Phenylephrine Related Compound E
  • Phenylephrine Related Compound F
  • Phenylephrine Related Compound G
  • Synonym(s):
  • 1-(3-Hydroxyphenyl)-2-(methylamino)ethan-1-one hydrochloride
  • (R)-3-{2-[Benzyl(methyl)amino]-1-hydroxyethyl}phenol
  • 2-[Benzyl(methyl)amino]-1-(3-hydroxyphenyl)ethan-1-one hydrochloride
  • (R)-2-Methyl-1,2,3,4-tetrahydroisoquinoline-4,8-diol hydrochloride monohydrate
  • (R)-N-(2-Hydroxy-2-(3-hydroxyphenyl)ethyl)-N-methylglycine
  • CAS No.:
  • 94240-17-2
  • 1367567-95-0
  • 71786-67-9
  • 1007885-60-0
  • 1094089-46-9
  • Formula:
  • C₉H₁₂ClNO₂
  • C₁₆H₁₉NO₂
  • C₁₆H₁₈ClNO₂
  • C₁₀H₁₃NO₂.HCl.H₂₀
  • C₁₁H₁₅NO₄

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1533024.
    Phenylephrine Related Compound C
    Phenylephrine Related Compound C
    USP reference standard
    Vial
    30 mg
    Log in for availability
    Log in for availability
    1 * 30 mg
    1 * 30 mg - 6 400 zł
    6 400 zł
    1 * 30 mg
       6 400 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1533024.
    Description
    Phenylephrine Related Compound C
    Synonym(s)
    1-(3-Hydroxyphenyl)-2-(methylamino)ethan-1-one hydrochloride
    CAS No.
    94240-17-2
    Formula
    C₉H₁₂ClNO₂
  • 1533035.
    Phenylephrine Related Compound D
    Phenylephrine Related Compound D
    USP reference standard
    Vial
    30 mg
    Log in for availability
    Log in for availability
    1 * 30 mg
    1 * 30 mg - 6 400 zł
    6 400 zł
    1 * 30 mg
       6 400 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1533035.
    Description
    Phenylephrine Related Compound D
    Synonym(s)
    (R)-3-{2-[Benzyl(methyl)amino]-1-hydroxyethyl}phenol
    CAS No.
    1367567-95-0
    Formula
    C₁₆H₁₉NO₂
  • 1533046.
    Phenylephrine Related Compound E
    Phenylephrine Related Compound E
    USP reference standard
    Vial
    30 mg
    Log in for availability
    Log in for availability
    1 * 30 mg
    1 * 30 mg - 6 340 zł
    6 340 zł
    1 * 30 mg
       6 340 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1533046.
    Description
    Phenylephrine Related Compound E
    Synonym(s)
    2-[Benzyl(methyl)amino]-1-(3-hydroxyphenyl)ethan-1-one hydrochloride
    CAS No.
    71786-67-9
    Formula
    C₁₆H₁₈ClNO₂
  • 1533261.
    Phenylephrine Related Compound F
    Phenylephrine Related Compound F
    USP reference standard
    Vial
    30 mg
    Log in for availability
    Log in for availability
    1 * 30 mg
    1 * 30 mg - 6 100 zł
    6 100 zł
    1 * 30 mg
       6 100 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1533261.
    Description
    Phenylephrine Related Compound F
    Synonym(s)
    (R)-2-Methyl-1,2,3,4-tetrahydroisoquinoline-4,8-diol hydrochloride monohydrate
    CAS No.
    1007885-60-0
    Formula
    C₁₀H₁₃NO₂.HCl.H₂₀
  • 1533272.
    Phenylephrine Related Compound G
    Phenylephrine Related Compound G
    USP reference standard
    Vial
    30 mg
    Log in for availability
    Log in for availability
    1 * 30 mg
    1 * 30 mg - 6 400 zł
    6 400 zł
    1 * 30 mg
       6 400 zł

    Specifications:
    More
    Specifications
    Cat. No.
    1533272.
    Description
    Phenylephrine Related Compound G
    Synonym(s)
    (R)-N-(2-Hydroxy-2-(3-hydroxyphenyl)ethyl)-N-methylglycine
    CAS No.
    1094089-46-9
    Formula
    C₁₁H₁₅NO₄
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.