Impurity and API standard, 5-(4'-Methylbiphenyl-2-yl)-1H-tetrazole
Catalog # PROOMM0168.05
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:50 mg
- Standards:Reference standards
- CAS No.:120568-11-8
- Description:5-(4'-Methylbiphenyl-2-yl)-1H-tetrazole
- Formula:C₁₄H₁₂N₄
- Description:5-(4'-Methylbiphenyl-2-yl)-1H-tetrazole