Order Entry
Poland
ContactUsLinkComponent
Organic reference standards, Sertraline hydrochloride assay standard, BP
Organic reference standards, Sertraline hydrochloride assay standard, BP
Certificates
Organic reference standards, Sertraline hydrochloride assay standard, BP

Specifications

  • CAS No.:
    79617-96-2
  • Formula:
    C₁₇H₁₈Cl₃N
  • Description:
    Sertraline hydrochloride impurity Standard

Specifications

Frequently Bought Together