Order Entry
Poland
ContactUsLinkComponent
Degreaser, Cen Mil E
Degreaser, Cen Mil E
Catalog # DIVY7516491
Supplier:  Diversey
Degreaser, Cen Mil E
Catalog # DIVY7516491
Supplier:  Diversey
Specifications
  • Description:
    Degreaser, Multi purpose
  • Packed:
    2×5 l containers/case
  • Pk:
    2 Can
  • pH value:
    12
Specifications
About this item
Multi-purpose degreaser that is ideal for routine applications.
  • Clear blue liquid
Can be used as a spray or with a sponge, pure or in solution in water.