Order Entry
Poland
ContactUsLinkComponent
 
Hydrogen peroxide 30% stabilised, Electronic Grade, J.T.Baker®
Catalog # 2204-01
Supplier:  Avantor
CAS Number:  
undefined
Hydrogen peroxide 30% stabilised, Electronic Grade, J.T.Baker®
Catalog # 2204-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1 pt US
  • Seq:
    8014

Specifications

Specification Test Results

  • For Microelectronic Use:
    30.0 - 32.0 %
  • Assay (H₂O₂):
    30.0 - 32.0 %
  • Color (APHA):
    ≤10
  • Free Acid (µeq/g):
    ≤0.2
  • Residue after Evaporation:
    ≤15 ppm
  • Ammonium (NH₄):
    ≤3 ppm
  • Chloride (Cl):
    ≤2 ppm
  • Nitrate (NO₃):
    ≤2 ppm
  • Phosphate (PO₄):
    ≤2 ppm
  • ACS - Sulfate (SO₄):
    ≤5 ppm
  • Trace Impurities - Aluminum (Al):
    ≤200.0 ppb
  • Trace Impurities - Antimony (Sb):
    ≤10.0 ppb
  • Trace Impurities - Arsenic (As):
    ≤10.0 ppb
  • Arsenic and Antimony (as As):
    ≤10.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤20.0 ppb
  • Trace Impurities - Beryllium (Be):
    ≤10.0 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤20.0 ppb
  • Trace Impurities - Boron (B):
    ≤10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤10.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤50.0 ppb
  • Trace Impurities - Chromium (Cr):
    ≤20.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤10.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤20.0 ppb
  • Trace Impurities - Germanium (Ge):
    ≤10.0 ppb
  • Trace Impurities - Gold (Au):
    ≤10.0 ppb
  • Heavy Metals (as Pb):
    ≤500.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤50.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤10.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤10.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤10.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤10.0 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤10.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤10.0 ppb
  • Trace Impurities - Niobium (Nb):
    ≤10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤600.0 ppb
  • Trace Impurities - Silicon (Si):
    ≤100.0 ppb
  • Trace Impurities - Silver (Ag):
    ≤10.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤500.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤10.0 ppb
  • Trace Impurities - Tantalum (Ta):
    ≤10.0 ppb
  • Trace Impurities - Thallium (Tl):
    ≤50.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤1000.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤10.0 ppb
  • Trace Impurities - Vanadium (V):
    ≤10.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤50 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤10.0 ppb

Specification Test Results

About this item

 :
Boiling Pt:∼107 °C (1013 hPa)
Melting Pt:<0 °C
Density:1.11 g/cm³ (20 °C)
ADR:5.1,II
 :34.01 g/mol
Storage Temperature:Ambient
EINECS:231-765-0
UN:2014
 :GHS03
 :GHS05
 :GHS06
 :GHS08
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333