Order Entry
Poland
ContactUsLinkComponent
 
Ammonia solution 29%, Finyte® for microelectronic, J.T.Baker®
Catalog # 5820-15
Supplier:  Avantor
CAS Number:  
undefined
Ammonia solution 29%, Finyte® for microelectronic, J.T.Baker®
Catalog # 5820-15
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    8 pt US
  • Seq:
    8026

Specifications

Specification Test Results

  • Appearance:
    Passes Test
  • Assay (as NH₃):
    28.0 - 30.0 %
  • Color (APHA):
    ≤10
  • Residue after Ignition:
    ≤3 ppm
  • Carbon Dioxide (CO₂):
    ≤10 ppm
  • Substances Reducing Permanganate:
    Passes Test
  • Chloride (Cl):
    ≤0.2 ppm
  • Nitrate (NO₃):
    ≤0.4 ppm
  • Phosphate (PO₄):
    ≤0.10 ppm
  • Total Sulfur (as SO₄):
    ≤0.2 ppm
  • Trace Impurities - Aluminum (Al):
    ≤10.0 ppb
  • Arsenic and Antimony (as As):
    ≤5.0 ppb
  • Trace Impurities - Barium (Ba):
    ≤10.0 ppb
  • Trace Impurities - Beryllium (Be):
    ≤10.0 ppb
  • Trace Impurities - Bismuth (Bi):
    ≤10.0 ppb
  • Trace Impurities - Boron (B):
    ≤10.0 ppb
  • Trace Impurities - Cadmium (Cd):
    ≤5.0 ppb
  • Trace Impurities - Calcium (Ca):
    ≤20 ppb
  • Trace Impurities - Chromium (Cr):
    ≤5.0 ppb
  • Trace Impurities - Cobalt (Co):
    ≤1.0 ppb
  • Trace Impurities - Copper (Cu):
    ≤10.0 ppb
  • Trace Impurities - Gallium (Ga):
    ≤10.0 ppb
  • Trace Impurities - Germanium (Ge):
    ≤10.0 ppb
  • Trace Impurities - Gold (Au):
    ≤10.0 ppb
  • Trace Impurities - Iron (Fe):
    ≤10.0 ppb
  • Trace Impurities - Lead (Pb):
    ≤10.0 ppb
  • Trace Impurities - Lithium (Li):
    ≤10.0 ppb
  • Trace Impurities - Magnesium (Mg):
    ≤10.0 ppb
  • Trace Impurities - Manganese (Mn):
    ≤5.0 ppb
  • Trace Impurities - Molybdenum (Mo):
    ≤10.0 ppb
  • Trace Impurities - Nickel (Ni):
    ≤5.0 ppb
  • Trace Impurities - Niobium (Nb):
    ≤10.0 ppb
  • Trace Impurities - Potassium (K):
    ≤10.0 ppb
  • Trace Impurities - Silicon (Si):
    ≤10.0 ppb
  • Trace Impurities - Silver (Ag):
    ≤5.0 ppb
  • Trace Impurities - Sodium (Na):
    ≤10.0 ppb
  • Trace Impurities - Strontium (Sr):
    ≤10.0 ppb
  • Trace Impurities - Tantalum (Ta):
    ≤10.0 ppb
  • Trace Impurities - Thallium (Tl):
    ≤10.0 ppb
  • Trace Impurities - Tin (Sn):
    ≤10.0 ppb
  • Trace Impurities - Titanium (Ti):
    ≤10.0 ppb
  • Trace Impurities - Vanadium (V):
    ≤10.0 ppb
  • Trace Impurities - Zinc (Zn):
    ≤10.0 ppb
  • Trace Impurities - Zirconium (Zr):
    ≤10.0 ppb
  • Particle Count - 0.5 µm and greater:
    ≤40 par/ml
  • Particle Count - 1.0 µm and greater:
    ≤10 par/ml

Specification Test Results

About this item

 :
 :35.05 g/mol
UN:2672
ADR:8,III
CAS Number:1336-21-6
 :7664-41-7
Formula:NH₃
MDL Number:MFCD00066650