Order Entry
Poland
ContactUsLinkComponent
Anti-VIL Mouse Monoclonal Antibody [clone: VIL1/1325]
Anti-VIL Mouse Monoclonal Antibody [clone: VIL1/1325]
Catalog # ABNOMAB15031
Supplier:  Abnova
Anti-VIL Mouse Monoclonal Antibody [clone: VIL1/1325]
Catalog # ABNOMAB15031
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Villin
  • Antigen symbol:
    VIL
  • Clonality:
    Monoclonal
  • Clone:
    VIL1/1325
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Storage buffer:
    In 10 mM PBS.
  • Sequence:
    MGPESTRMERLRGMTLAKEIRDQERGGRTYVGVVDGENELASPKLMEVMNHVLGKRRELKAAVPDTVVEPALKAALKLYHVSDSEGNLVVREVATRPLTQDLLSHEDCYILDQGGLKIYVWKGKKANEQEKKGA
  • Storage temperature:
    Store at -20 to –80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to 133 residues of human Villin.
  • Purification:
    Protein A/G purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant human Villin.

Type: Primary
Antigen: VIL
Clonality: Monoclonal
Clone: VIL1/1325
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human