Order Entry
Poland
ContactUsLinkComponent
Anti-UBE2G2 Rabbit Polyclonal Antibody
Anti-UBE2G2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB30852
Supplier:  Abnova
Anti-UBE2G2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB30852
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    UBE2G2
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    7327
  • Storage buffer:
    In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
  • Sequence:
    LSFPLDYPLSPPKMRFTCEMFHPNIYPDGRVCISILHAPGDDPMGYESSAERWSPVQSVEKILLSVVSMLAEPNDESGANVDASKMWRDDREQFYKIAKQIVQKS
  • Storage temperature:
    Store at 4 °C. For long term storage store at -20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human UBE2G2.
  • Purification:
    Affinity purification
  • Size:
    100 µL
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against partial recombinant human UBE2G2.

Type: Primary
Antigen: UBE2G2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human