Order Entry
Poland
ContactUsLinkComponent
Anti-TFRC Mouse Monoclonal Antibody [clone: TFRC/1818]
Anti-TFRC Mouse Monoclonal Antibody [clone: TFRC/1818]
Catalog # ABNOMAB15004
Supplier:  Abnova
Anti-TFRC Mouse Monoclonal Antibody [clone: TFRC/1818]
Catalog # ABNOMAB15004
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TFRC
  • Clonality:
    Monoclonal
  • Clone:
    TFRC/1818
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    7037
  • Storage buffer:
    In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
  • Storage temperature:
    Store at 4 °C.
  • Immunogen:
    Recombinant protein corresponding to amino acids 94-212 of human TFRC.
  • Purification:
    Protein A/G purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against partial recombinant human TFRC.

Type: Primary
Antigen: TFRC
Clonality: Monoclonal
Clone: TFRC/1818
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human