Anti-SOX2 Mouse Monoclonal Antibody [clone: SOX2/1792]
Catalog # ABNOMAB14989
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:SOX2
- Clonality:Monoclonal
- Clone:SOX2/1792
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:6657
- Storage buffer:In 10 mM PBS.
- Storage temperature:Store at -20 to –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 176-305 of human SOX2.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant human SOX2.
Type: Primary
Antigen: SOX2
Clonality: Monoclonal
Clone: SOX2/1792
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human