Anti-SLC3A2 Mouse Monoclonal Antibody (PerCP) [clone: FG1/8]
Catalog # ABNOMAB15378
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:4F2 cell-surface antigen heavy chain
- Antigen symbol:SLC3A2
- Clonality:Monoclonal
- Clone:FG1/8
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:6520
- Storage buffer:In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
- Storage temperature:Store in the dark at 4 °C.
- Immunogen:T cells from leukemic HPB-ALL.
- Purification:Affinity purification
- Size:100 AS
- Pk:100 Assays
Specifications
About this item
Mouse monoclonal antibody raised against human SLC3A2.
Type: Primary
Antigen: SLC3A2
Clonality: Monoclonal
Clone: FG1/8
Conjugation: PerCP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human