Anti-MAML2 Mouse Monoclonal Antibody [clone: MAML2/1302]
Catalog # ABNOMAB15049
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:MAML2
- Clonality:Monoclonal
- Clone:MAML2/1302
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Form:Liquid
- Gene ID:84441
- Storage buffer:In 10 mM PBS.
- Storage temperature:Store at -20 to –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human MAML2.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant human MAML2.
Type: Primary
Antigen: MAML2
Clonality: Monoclonal
Clone: MAML2/1302
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human