Anti-Ly6a Rat Monoclonal Antibody (CF-Blue) [clone: D7]
Catalog # ABNOMAB15528
Dostawca: Abnova
Dane techniczne
- Antibody type:Primary
- Antigen name:Lymphocyte antigen 6 complex, locus A
- Antigen symbol:Ly6a
- Clonality:Monoclonal
- Clone:D7
- Conjugation:CF-Blue™
- Host:Rat
- Isotype:IgG2a
- Reactivity:Mouse
- Form:Liquid
- Gene ID:110454
- Storage buffer:In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
- Storage temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light.
- Immunogen:IL-2-dependent mouse T-cell line CTL-L.
- Purification:Affinity purification
- Size:100 µg
- Pk:100 µG
Dane techniczne
Informacje o tym produkcie
Rat monoclonal antibody raised against native mouse Ly6a.
Type: Primary
Antigen: Ly6a
Clonality: Monoclonal
Clone: D7
Conjugation: CF-Blue
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse