Anti-KRT10 Mouse Monoclonal Antibody [clone: SPM623]
ABNOMAB14832
: Abnova
- Antibody type:Primary
- Antigen symbol:KRT10
- Clonality:Monoclonal
- Clone:SPM623
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:3858
- Storage buffer:In 10 mM PBS.
- Storage temperature:Store at -20 to –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Cytoskeletal preparation extracted from human ectocervical epithelium.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Mouse monoclonal antibody raised against native human KRT10.
Type: Primary
Antigen: KRT10
Clonality: Monoclonal
Clone: SPM623
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human