Anti-INSL3 Rabbit Polyclonal Antibody
ABNOPAB30995
: Abnova
- Antibody type:Primary
- Antigen symbol:INSL3
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:3640
- Storage buffer:In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
- Sequence:EMREKLCGHHFVRALVRVCGGPRWSTEARRPATGGDRELLQWLERRHLLHGLVADSNLTLGPGLQPLPQTSHHHRHHRAAATNPARYCCLSGCTQQDLLTLCPY
- Storage temperature:Store at 4 °C. For long term storage store at -20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human INSL3.
- Purification:Antigen affinity purification
- Size:100 µL
- Pk:100 µl
Rabbit polyclonal antibody raised against partial recombinant human INSL3.
Type: Primary
Antigen: INSL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human