Anti-FOXA1 Mouse Monoclonal Antibody [clone: FOXA1/1515]
Catalog # ABNOMAB14813
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen symbol:FOXA1
- Clonality:Monoclonal
- Clone:FOXA1/1515
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:3169
- Amino acid number:467-614 aa
- Storage buffer:In 10 mM PBS.
- Storage temperature:Store at -20 to –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 467-614 of human FOXA1.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant human FOXA1.
Type: Primary
Antigen: FOXA1
Clonality: Monoclonal
Clone: FOXA1/1515
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human