Anti-ENG Mouse Monoclonal Antibody (PE) [clone: 2H6F11]
ABNOMAB15389
: Abnova
- Antibody type:Primary
- Antigen name:Endoglin
- Antigen symbol:ENG
- Clonality:Monoclonal
- Clone:2H6F11
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:2022
- Storage buffer:In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
- Storage temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light.
- Immunogen:Recombinant protein corresponding to human ENG.
- Purification:Affinity purification
- Size:100 AS
- Pk:100 Assays
Mouse monoclonal antibody raised against recombinant human ENG.
Type: Primary
Antigen: ENG
Clonality: Monoclonal
Clone: 2H6F11
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human