Order Entry
Poland
ContactUsLinkComponent
 
Anti-CHGA Rabbit Polyclonal Antibody
  ABNOPAB30816
 :  Abnova
Anti-CHGA Rabbit Polyclonal Antibody
  ABNOPAB30816
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Chromogranin A
  • Antigen symbol:
    CHGA
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    1113
  • Storage buffer:
    In 10 mM PBS.
  • Storage temperature:
    Store at -20 to –80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to full length human CHGA.
  • Purification:
    Protein A purification
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody raised against full length recombinant human CHGA.

Type: Primary
Antigen: CHGA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG, kappa
Reactivity: Human