Anti-CDH2 Mouse Monoclonal Antibody [clone: CDH2/1573]
ABNOMAB14718
: Abnova
- Antibody type:Primary
- Antigen symbol:CDH2
- Clonality:Monoclonal
- Clone:CDH2/1573
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Form:Liquid
- Gene ID:1000
- Storage buffer:In 10 mM PBS.
- Storage temperature:Store at -20 to –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to intracellular domain of human CDH2.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Mouse monoclonal antibody raised against partial recombinant human CDH2.
Type: Primary
Antigen: CDH2
Clonality: Monoclonal
Clone: CDH2/1573
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Mouse