Order Entry
Poland
ContactUsLinkComponent
 
Anti-CD80 Hamster Monoclonal Antibody (FITC) [clone: 16-10A1]
  ABNOMAB15457
 :  Abnova
Anti-CD80 Hamster Monoclonal Antibody (FITC) [clone: 16-10A1]
  ABNOMAB15457
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    CD80 molecule
  • Antigen symbol:
    CD80
  • Clonality:
    Monoclonal
  • Clone:
    16-10A1
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Hamster
  • Isotype:
    IgG2
  • Reactivity:
    Mouse
  • Form:
    Liquid
  • Gene ID:
    12519
  • Storage buffer:
    In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
  • Storage temperature:
    Store in the dark at 4 °C. Avoid prolonged exposure to light.
  • Immunogen:
    Mouse Cd80 (B7) transfected cell line.
  • Purification:
    Affinity purification
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Hamster monoclonal antibody raised against native mouse Cd80.

Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: 16-10A1
Conjugation: FITC
Epitope:
Host: Hamster
Isotype: IgG2
Reactivity: Mouse