Anti-CD3e Hamster Monoclonal Antibody (CF-Blue) [clone: 145-2C11]
Catalog # ABNOMAB15456
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD3e molecule
- Antigen symbol:CD3e
- Clonality:Monoclonal
- Clone:145-2C11
- Conjugation:CF-Blue™
- Host:Hamster
- Isotype:IgG
- Reactivity:Mouse
- Form:Liquid
- Gene ID:12501
- Storage buffer:In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
- Storage temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light.
- Immunogen:H-2Kb specific cytotoxic T lymphocyte clone BM10-37.
- Purification:Affinity purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Hamster monoclonal antibody raised against native mouse Cd3e.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: 145-2C11
Conjugation: CF-Blue
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse