Order Entry
Poland
ContactUsLinkComponent
Anti-CD1A Mouse Monoclonal Antibody [clone: CBT6]
Anti-CD1A Mouse Monoclonal Antibody [clone: CBT6]
Catalog # ABNOMAB15053
Dostawca:  Abnova
Anti-CD1A Mouse Monoclonal Antibody [clone: CBT6]
Catalog # ABNOMAB15053
Dostawca:  Abnova

Dane techniczne

  • Antibody type:
    Primary
  • Antigen symbol:
    CD1A
  • Clonality:
    Monoclonal
  • Clone:
    CBT6
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    909
  • Storage buffer:
    In 10 mM PBS.
  • Storage temperature:
    Store at -20 to –80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified human CD1A.
  • Purification:
    Protein A/G purification
  • Size:
    100 µg
  • Pk:
    100 µG

Dane techniczne

Informacje o tym produkcie

Mouse monoclonal antibody raised against native human CD1A.

Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: CBT6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human