Order Entry
Poland
ContactUsLinkComponent
Polyethyleneimine, M.W. ~1300 (LS) 50% in water, Sigma-Aldrich®
Polyethyleneimine, M.W. ~1300 (LS) 50% in water, Sigma-Aldrich®
Supplier:  Merck
Certificates
Polyethyleneimine, M.W. ~1300 (LS) 50% in water, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Appearance (Colour):
    Colorless to yellow
  • Appearance (Form):
    Viscous liquid
  • Infrared spectrum:
    Conforms to Structure
  • Concentration:
    48,0 - 52,0%
  • Viscosity sec (flow cup):
    40 - 100
  • Viscosity:
    200 - 500 cP (25 °C) (lit.)

Specification Test Results

Frequently Bought Together