Specifications
- Antibody type:Primary
- Antigen name:DEAH (Asp-Glu-Ala-His) box polypeptide 32
- Antigen symbol:DHX32
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:DHLP1|DDX32
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
DEAD-box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp, are putative RNA helicases implicated in several cellular processes involving modifications of RNA secondary structure and ribosome/spliceosome assembly. Based on their distribution patterns, some members of this family may be involved in embryogenesis, spermatogenesis, and cellular growth and division. DDX32 is a 743 amino acid nuclear protein that localizes to the mitochondria and is a member of the DEAD box helicase family. Expressed in various tissues, DDX32 is up-regulated by ionomycin in T lymphocytes and down-regulated in acute lymphoblastic leukemia. Considered a novel RNA helicase, DDX32 may play an important role in the development of colorectal cancer and may be involved in regulating T-cell response to certain apoptotic stimuli.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DHX32
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat