Anti-HCLS1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-5109R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:hematopoietic cell-specific Lyn substrate 1
- Antigen symbol:HCLS1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:3059
- Antigen synonyms:HS1|p75|CTTNL|lckBP1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:200-250/486
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Substrate of the antigen receptor-coupled tyrosine kinase. Plays a role in antigen receptor signaling for both clonal expansion and deletion in lymphoid cells. May also be involved in the regulation of gene expression.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HCLS1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat