Order Entry
Poland
ContactUsLinkComponent
 
Anti-RUNX3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
  BOSSBS-4848R-A488
 :  Bioss
Anti-RUNX3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
  BOSSBS-4848R-A488
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Runt-related Transcription Factor 3
  • Antigen symbol:
    RUNX3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    864
  • Antigen synonyms:
    AML2|Core-binding factor subunit alpha-3|RUNX3|SL3-3 enhancer factor 1 alpha C subunit|PEBP2A3|Runt-related transcription factor 3|Oncogene AML-2|CBFA3|Polyomavirus enhancer-binding protein 2 alpha C subunit|Acute myeloid leukemia 2 protein|SL3/
  • Modification:
    phospho-Ser338
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

RUNX3 binds to the core site of murine Leukemia virus, the core sequences in the enhancer of the polyomavirus, and also to the enhancers of the T-cell receptor genes. May be involved in the control of cellular proliferation and/or differentiation (By similarity). Heterodimer of an alpha and a beta subunit. The alpha subunit binds DNA as a monomer and through the Runt domain.DNA-binding is increased by heterodimerization. Interacts with TLE1 and SUV39H1, Subcellular location in Nucleus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RUNX3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human