Anti-CSF3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-1023R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Colony Stimulating Factor 3 (granulocyte)
- Antigen symbol:CSF3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1440
- Antigen synonyms:C17orf33|GCSF|CSF3OS
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:176-207/207
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Granulocyte/macrophage colony-stimulating factors are cytokines that act in hematopoiesis by controlling the production, differentiation, and function of 2 related white cell populations of the blood, the granulocytes and the monocytes-macrophages. This CSF induces granulocytes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CSF3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity: