Order Entry
Poland
ContactUsLinkComponent
Anti-FOSL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-FOSL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
  BOSSBS-3158R-A488
 :  Bioss
Anti-FOSL1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
  BOSSBS-3158R-A488
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    FOS-Like Antigen 1
  • Antigen symbol:
    FOSL1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    8061
  • Antigen synonyms:
    FRA1|fra-1|FRA
  • Modification:
    phospho-Ser265
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    230-271/271
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. [provided by RefSeq].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FRA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat