Order Entry
Poland
ContactUsLinkComponent
Anti-AQP9 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anti-AQP9 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-2060R-A555
Supplier:  Bioss
Anti-AQP9 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-2060R-A555
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Aquaporin 9
  • Antigen symbol:
    AQP9
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    366
  • Antigen synonyms:
    HsT17287|SSC1|AQP-9
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    211-246/295
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Forms a channel with a broad specificity. Mediates passage of a wide variety of non-charged solutes including carbamides, polyols, purines, and pyrimidines in a phloretin- and mercury-sensitive manner, whereas amino acids, cyclic sugars, Na(+), K(+), Cl(-), and deprotonated monocarboxylates are excluded. Also permeable to urea and glycerol.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: AQP9
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat