Order Entry
Poland
ContactUsLinkComponent
Anti-ECE2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-ECE2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-1753R-A488
Supplier:  Bioss
Anti-ECE2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-1753R-A488
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Endothelin Converting Enzyme 2
  • Antigen symbol:
    ECE2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 488
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    9718
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    780-830/883
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Converts big endothelin-1 to endothelin-1. Also involved in the processing of various neuroendocrine peptides, including neurotensin, angiotensin I, substance P, proenkephalin-derived peptides, and prodynorphin-derived peptides. May limit beta-amyloid peptide accumulation in brain. May also have methyltransferase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ECE2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat