Order Entry
Poland
ContactUsLinkComponent
Anti-CCNF Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-CCNF Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-1686R-A350
 :  Bioss
Anti-CCNF Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-1686R-A350
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin F
  • Antigen symbol:
    CCNF
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    899
  • Antigen synonyms:
    FBX1|FBXO1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    480-530/786
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Substrate recognition component of a SCF (SKP1-CUL1-F-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of CP110 during G2 phase, thereby acting as an inhibitor of centrosome reduplication.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCNF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat