Specifications
- Antibody type:Primary
- Antigen name:Apolipoprotein L5
- Antigen symbol:APOL5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:80831
- Antigen synonyms:APOLV|APOL-V
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The apolipoprotein L gene family maps to a region on chromosome 22 and encodes six highly homologous proteins designated apoL-I, apoL-II, apoL-III, apoL-IV, apoL-V and apoL-VI, all of which function as components of plasma lipoproteins. ApoL-V (apolipoprotein L-V), also known as APOL5, is a 433 amino acid protein that localizes to the cytoplasm and belongs to the apolipoprotein L family. Expressed in a variety of tissues including testis, stomach, uterus and skeletal muscle, apoL-V is thought to affect the movement of lipids in the cytoplasm and may allow the binding of lipids to organelles. Like other members of the apolipoprotein L family, apoL-V is thought to be involved in the development of schizophrenia.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: APOL5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human