Specifications
- Antibody type:Primary
- Antigen name:WNT1 Inducible Signaling Pathway Protein 3
- Antigen symbol:WISP3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:8838
- Antigen synonyms:CCN6|WNT1-inducible-signaling pathway protein 3|WISP-3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Wnt-induced secreted protein (WISP)-1, WISP-2 and WISP-3 are members of the CCN family of growth factors, which include connective tissue growth factor (CTGF) and Cyr61. WISP-1, WISP-2 and WISP-3 share significant sequence similarity, including four conserved cysteine-rich domains, and they are believed to function as dimers in their active forms. WISP-1 expression is observed in various tissues including adult heart, kidney and spleen, while WISP-2 expression predominates in skeletal muscle, colon and ovary. Both WISP-1 and WISP-2 are upregulated in cells transformed with the proto-oncogene Wnt-1, and they are also more highly expressed in human colon tumors, suggesting that these proteins may participate in tumor development. WISP-3 is involved in normal post-natal skeletal growth, and it is also implicated in the development of the autosomal recessive skeletal disorder progressive pseudorheumatoid dysplasia, which affects cartilage homeostasis by disrupting the growth of chondrocyte and normal cell columnar organization.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: WISP3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat