- Antibody type:Primary
- Antigen name:Alveolar Soft Part Sarcoma Chromosome Region, Candidate 1
- Antigen symbol:ASPSCR1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:79058
- Antigen synonyms:ASPL|TUG|RCC17|ASPCR1|UBXD9|ASPS|UBXN9
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Glut4 is a twelve pass transmembrane protein (12TM) whose carboxy-terminus may dictate its cellular localization. Aberrant Glut4 expression has been suggested to contribute to such maladies as obesity and diabetes. Glut4 null mice have shown that while functional Glut4 protein is not required for maintaining normal glucose levels, it is necessary for sustained growth, normal cellular glucose, fat metabolism and prolonged longevity. TUG (ASPL in humans) regulates the trafficking of glucose via Glut4. Full-length TUG forms a complex with Glut4 and in 3T3-L1 adipocytes and this complex is present in unstimulated cells and is disassembled by insulin. TUG acts by trapping endocytosed Glut4 and tethering it intracellularly. Insulin mobilizes this pool of retained Glut4 by releasing the tether.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ASPSCR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat