Specifications
- Antibody type:Primary
- Antigen name:Testis specic kinase substrate
- Antigen symbol:TSKS
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:60385
- Antigen synonyms:TSKS_HUMAN.|TSKS|Testis specic kinase substrate|STK22 substrate 1|Stk22s1|Testis specic serine kinase substrate|MGC134104|TSKS1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
TSKS is a 592 amino acid protein that is highly expressed in human testicular tissue. Low levels of TSKS are detectable in prostate, placenta, fetal liver, thymus, and mammary gland tissues. TSKS is found to be downregulated in cancerous testicular tissue from seminoma, teratocarcinoma, embryonal and Leydig cell tumors concurrently with high expression in neighboring premalignant carcinoma. TSKS protein contains an N-terminal signal peptide, but does not contain a transmembrane region. TSKS has many potential phosphorylation and glycosylation sites and is phosphorylated by soluble recombinant TSSK2 in vitro. It is thought that TSKS likely plays a physiological role in spermatogenesis or spermiogenesis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TSKS
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat