- Antibody type:Primary
- Antigen name:Prostaglandin I2 Receptor
- Antigen symbol:PTGIR
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:PI2R_HUMAN|Prostaglandin I2 receptor|PGI receptor|PTGIR.|Prostacyclin receptor|IP|PGI2 receptor|Prostanoid IP receptor|PRIPR
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Cyclooxygenases metabolize arachidonate to five primary prostanoids: PGE2, PGF2?, PGI2, TXA2 and PGD2. These lipid mediators interact with specific members of G protein-coupled prostanoid receptors, designated EP, FP, IP, TP and DP, respectively. The IP Receptor binds prostacyclin, PGI2, the main pro-stanoid synthesized by vascular tissues.Upon binding to the IP Receptor, prostacyclin activates adenylate cyclase primarily through the Gas protein. The gene encoding the human IP Receptor is located on chromosome 19. It is expressed as a glycosylated and phosphorylated protein, which is abundantly expressed in vascular tissues such as aorta, lung, atrium and ventricle, as well as in kidney, thymus, spleen and neurons.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PTGIR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat