Specifications
- Antibody type:Primary
- Antigen name:Caspase 4 And Caspase 13
- Antigen symbol:CASP4 + CASP 13
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Bovine
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Caspase 13 subunit 2|CASP-13|apoptosis related cysteine|Caspase-13|ERICE.|CASPD_BOVIN|Caspase 4|Caspase 13 subunit 1|CASP4|Caspase 4 subunit 1|Evolutionary related interleukin-1-beta-converting enzyme
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Caspase-13 (also designated Evolutionarily related interleukin-1beta converting enzyme ; ERICE) involved in the activation cascade of caspases responsible for apoptosis execution. Might function by either activating some proteins required for cell death or inactivating proteins necessary for cell survival. [subunit] Heterodimer of a small and a large subunit. The two subunits are derived from the precursor sequence by an autocatalytic mechanism or by cleavage by Caspase-8. Mainly expressed in peripheral blood lymphocytes, spleen and placenta. Belongs to peptidase family C14.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CASP4 + CASP 13
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Bovine