Order Entry
Poland
ContactUsLinkComponent
Acetonitrile with 0.1% (v/v) trifluoroacetic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
 
 
Acetonitrile with 0.1% (v/v) trifluoroacetic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
  1.59014.2500
 :  Merck
 :  
 
 
Acetonitrile with 0.1% (v/v) trifluoroacetic acid, LiChrosolv®, hypergrade for LC-MS, Supelco®
  1.59014.2500
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Acid content:
    0.095 - 0.105 % (v/v)
  • Identity:
    passes test
  • Color:
    ≤ 10 Hazen
  • Al (Aluminium):
    ≤ 0.05 ppm
  • Ca (Calcium):
    ≤ 0.05 ppm
  • Fe (Iron):
    ≤ 0.05 ppm
  • K (Potassium):
    ≤ 0.025 ppm
  • Mg (Magnesium):
    ≤ 0.05 ppm
  • Na (Sodium):
    ≤ 0.25 ppm
  • Gradient grade (at 210 nm):
    ≤ 1.0 mAU
  • Gradient grade (at 254 nm):
    ≤ 0.5 mAU
  • Suitable for LC-MS; Intensity of background mass peak based on reserpine (APCI/ESI positive):
    ≤ 2 ppb

 

 

 :
 :0.786 g/cm3 (20 °C)
 :2 °C
 :3,
 :M-Clarity™ quality level = MQ100
 :1648
 :GHS02
 :GHS07
 :Ambient
 :75-05-8