Anti-MMP-11 Rabbit Polyclonal Antibody [clone: G88]
Catalog # BWRLBS1230
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:MMP-11
- Clonality:Polyclonal
- Clone:G88
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 55-100 of Human MMP-11.
MMP-11 (G88) pAb detects endogenous levels of MMP-11 protein.
Type: Primary
Antigen: MMP-11
Clonality: Polyclonal
Clone: G88
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat