Anti-Histone H3 Rabbit Polyclonal Antibody [clone: Tri-Methyl K14]
Catalog # BWRLBS7525
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Histone H3
- Clonality:Polyclonal
- Clone:Tri-Methyl K14
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide conjugated to KLH derived from within residues 1-100 of Human Histone H3, Tri methylated at K14.
Histone H3 (Tri-Methyl K14) pAb detects endogenous levels of histone H3 only when Tri-methylated at Lys14.
Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone: Tri-Methyl K14
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat; Other (Wide Range)