Anti-DLC1 Rabbit Polyclonal Antibody
100 µg
Catalog # BWRLBS7318
Supplier: Bioworld Technology
Jump to:
Product Details & Documents
Recombinant full length Human DLC1.
DLC1 pAb detects endogenous levels of DLC1 protein.
Type: Primary
Antigen: DLC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Specifications
- Size:100 µg
- Western blot:Yes
- Cat. No.:BWRLBS7318
- Clonality:Polyclonal
- Reactivity:Human|Mouse
- Antibody type:Primary
- Host:Rabbit
- ImmunoChemistry:Yes
- Antigen symbol:DLC1
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



